Assay Details
Target Gene Details
Entrez Gene ID: | 57514 |
Gene Name: | Rho GTPase activating protein 31 |
Gene Aliases: |
AOS1, CDGAP |
Location: |
Chr.3:119294289-119419476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP31 | NM_020754.3 | NP_065805.2 | ||
| XM_006713714.3 | XP_006713777.1 | |||
| XM_017006955.1 | XP_016862444.1 | |||
| AB033030.1 | ||||
| AK293726.1 | ||||
| BC112163.1 | AAI12164.1 | |||
| BC112165.1 | AAI12166.1 |
Target Gene Details
Entrez Gene ID: | 100874246 |
Gene Name: | ARHGAP31 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:119314293-119322760 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP31-AS1 | NR_046748.1 | |||
| DA269443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829694 | Chr.3:119292641 - 119434732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map