Assay Details
Target Gene Details
Entrez Gene ID: | 6138 |
Gene Name: | ribosomal protein L15 |
Gene Aliases: |
DBA12, EC45, L15, RPL10, RPLY10, RPYL10 |
Location: |
Chr.3:23916804-23923696 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL15 | NM_001253379.1 | NP_001240308.1 | ||
| NM_001253380.1 | NP_001240309.1 | |||
| NM_001253382.1 | NP_001240311.1 | |||
| NM_001253383.1 | NP_001240312.1 | |||
| NM_001253384.1 | NP_001240313.1 | |||
| NM_002948.3 | NP_002939.2 | |||
| AF279903.1 | AAG44837.1 | |||
| AF283772.2 | AAG15591.1 | |||
| AK098795.1 | ||||
| BC014837.1 | AAH14837.1 | |||
| BC081565.1 | AAH81565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map