Assay Details
Target Gene Details
Entrez Gene ID: | 23395 |
Gene Name: | leucyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
LEURS, PRLTS4, mtLeuRS |
Location: |
Chr.3:45388561-45548842 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LARS2 | NM_015340.3 | NP_056155.1 | ||
XM_005265006.1 | XP_005265063.1 | |||
XM_011533554.2 | XP_011531856.1 | |||
XM_017006042.1 | XP_016861531.1 | |||
AK308146.1 | ||||
BC025989.2 | AAH25989.1 | |||
BI463427.1 | ||||
D21851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834677 | Chr.3:45343270 - 45511215 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
