Assay Details
Target Gene Details
Entrez Gene ID: | 29945 |
Gene Name: | anaphase promoting complex subunit 4 |
Gene Aliases: |
APC4 |
Location: |
Chr.4:25377226-25418498 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC4 | NM_001286756.1 | NP_001273685.1 | ||
| NM_013367.2 | NP_037499.2 | |||
| XM_005248159.2 | XP_005248216.1 | |||
| XM_011513838.1 | XP_011512140.1 | |||
| XM_011513839.1 | XP_011512141.1 | |||
| XM_017008138.1 | XP_016863627.1 | |||
| AF191338.1 | AAF05752.1 | |||
| AK023597.1 | ||||
| AK292336.1 | ||||
| BC059383.1 | AAH59383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997544 | Chr.4:25362237 - 25915432 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77801920] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map