Assay Details
Target Gene Details
Entrez Gene ID: | 22998 |
Gene Name: | LIM and calponin homology domains 1 |
Gene Aliases: |
LIMCH1A, LMO7B |
Location: |
Chr.4:41359607-41700044 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIMCH1 | NM_001289124.1 | NP_001276053.1 | ||
| CR936610.1 | CAI56754.1 |
More Information
Set Membership: |
|

Genomic Map