Assay Details
Target Gene Details
Entrez Gene ID: | 550112 |
Gene Name: | UBA6 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:67701278-67722505 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBA6-AS1 | NR_015439.1 | |||
AK093682.1 | ||||
AK129519.1 | ||||
DA770515.1 |
