Assay Details
Target Gene Details
Entrez Gene ID: | 10563 |
Gene Name: | C-X-C motif chemokine ligand 13 |
Gene Aliases: |
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13 |
Location: |
Chr.4:77511753-77611834 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCL13 | NM_006419.2 | NP_006410.1 | ||
AF044197.1 | AAC14402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3601076 | Chr.4:77529554 - 77536517 on Build GRCh38 | Loss |
![]() |
nsv829975 | Chr.4:77505056 - 77644044 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
