Assay Details
Target Gene Details
Entrez Gene ID: | 51138 |
Gene Name: | COP9 signalosome subunit 4 |
Gene Aliases: |
CSN4 |
Location: |
Chr.4:83035086-83075818 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS4 | NM_001258006.1 | NP_001244935.1 | ||
| NM_016129.2 | NP_057213.2 | |||
| XM_011532019.1 | XP_011530321.1 | |||
| AF100757.1 | AAD43021.1 | |||
| AK001148.1 | ||||
| AK001210.1 | BAA91555.1 | |||
| AK024005.1 | ||||
| AK094238.1 | ||||
| AK223178.1 | BAD96898.1 | |||
| BC004302.1 | AAH04302.1 | |||
| BC009292.2 | AAH09292.1 | |||
| BC093007.1 | AAH93007.1 | |||
| KU178633.1 | ||||
| KU178634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829990 | Chr.4:83035297 - 83188940 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map