Assay Details
Target Gene Details
Entrez Gene ID: | 116984 |
Gene Name: | ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
CENTD1, PARX |
Location: |
Chr.4:36005186-36244832 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARAP2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829905 | Chr.4:35895023 - 36085836 on Build GRCh38 | Gain |
|
| esv3569373 | Chr.4:35669991 - 36407345 on Build GRCh38 | Loss |
|

Genomic Map