Assay Details
Target Gene Details
Entrez Gene ID: | 3381 |
Gene Name: | integrin binding sialoprotein |
Gene Aliases: |
BNSP, BSP, BSP-II, SP-II |
Location: |
Chr.4:87799550-87812449 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IBSP | NM_004967.3 | NP_004958.2 | ||
| AI751848.1 | ||||
| J05213.1 | AAC95490.1 |
More Information
Set Membership: |
|

Genomic Map