Assay Details
Target Gene Details
Entrez Gene ID: | 57630 |
Gene Name: | SH3 domain containing ring finger 1 |
Gene Aliases: |
POSH, RNF142, SH3MD2 |
Location: |
Chr.4:169094256-169271098 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF1 | NM_020870.3 | NP_065921.2 | ||
| AB040927.1 | BAA96018.1 | |||
| AK025255.1 | ||||
| BC053671.1 | AAH53671.1 |
More Information
Set Membership: |
|

Genomic Map