Assay Details
Target Gene Details
Entrez Gene ID: | 7345 |
Gene Name: | ubiquitin C-terminal hydrolase L1 |
Gene Aliases: |
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1 |
Location: |
Chr.4:41256881-41268429 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCHL1 | NM_004181.4 | NP_004172.2 | ||
| AB209038.1 | 1 | 5591 | BAD92275.1 | |
| AK054579.1 | ||||
| AK315368.1 | ||||
| BC000332.2 | AAH00332.1 | |||
| BC005117.1 | AAH05117.1 | |||
| BC006305.2 | AAH06305.1 | |||
| FJ224355.1 | ||||
| GQ472201.1 | ||||
| X04741.1 | CAA28443.1 |
More Information
Set Membership: |
|

Genomic Map