Assay Details
Target Gene Details
Entrez Gene ID: | 8821 |
Gene Name: | inositol polyphosphate-4-phosphatase type II B |
Gene Aliases: |
- |
Location: |
Chr.4:142023160-142846535 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP4B | NM_001101669.1 | NP_001095139.1 | ||
| NM_003866.2 | NP_003857.2 | |||
| XM_005263323.3 | XP_005263380.1 | |||
| XM_005263325.3 | XP_005263382.1 | |||
| XM_011532391.2 | XP_011530693.1 | |||
| XM_011532392.2 | XP_011530694.1 | |||
| XM_017008794.1 | XP_016864283.1 | |||
| XM_017008795.1 | XP_016864284.1 | |||
| XM_017008796.1 | XP_016864285.1 | |||
| XM_017008797.1 | XP_016864286.1 | |||
| XM_017008798.1 | XP_016864287.1 | |||
| BC017924.2 | AAH17924.1 | |||
| BC110918.1 | AAI10919.1 | |||
| BC133005.1 | ||||
| BX649090.1 | ||||
| U96922.1 | AAB72153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830097 | Chr.4:142028552 - 142215219 on Build GRCh38 | Gain |
|
| nsv1027781 | Chr.4:142022332 - 142385414 on Build GRCh38 | Gain |
|
| nsv830096 | Chr.4:141945946 - 142102608 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map