Assay Details
Target Gene Details
Entrez Gene ID: | 442117 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase-like 6 |
Gene Aliases: |
GALNACT20, GALNT17, GalNAc-T6L |
Location: |
Chr.4:171812690-173040408 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNTL6 | NM_001034845.2 | NP_001030017.2 | ||
| XM_011531993.2 | XP_011530295.1 | |||
| XM_017008243.1 | XP_016863732.1 | |||
| XM_017008244.1 | XP_016863733.1 | |||
| AJ626725.1 | CAF25036.1 | |||
| AK295464.1 | ||||
| BC044793.1 | ||||
| BC047551.1 | AAH47551.1 | |||
| GU220060.1 |
Target Gene Details
Entrez Gene ID: | 101928314 |
Gene Name: | uncharacterized LOC101928314 |
Gene Aliases: |
- |
Location: |
Chr.4:172629932-172726878 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928314 | NR_125895.1 | |||
| BI461914.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs80291412] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map