Assay Details
Target Gene Details
Entrez Gene ID: | 23321 |
Gene Name: | tripartite motif containing 2 |
Gene Aliases: |
CMT2R, RNF86 |
Location: |
Chr.4:153152281-153339322 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM2 | NM_001130067.1 | NP_001123539.1 | ||
| XM_011531796.2 | XP_011530098.1 | |||
| XM_011531798.2 | XP_011530100.1 | |||
| XM_017007948.1 | XP_016863437.1 | |||
| XM_017007951.1 | XP_016863440.1 | |||
| XM_017007952.1 | XP_016863441.1 | |||
| BC011052.1 | AAH11052.1 |
More Information
Set Membership: |
|

Genomic Map