Assay Details
Target Gene Details
Entrez Gene ID: | 93627 |
Gene Name: | TBC1 domain containing kinase |
Gene Aliases: |
HSPC302, IHPRF3, TBCKL |
Location: |
Chr.4:106044317-106316704 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCK | NM_001163435.2 | NP_001156907.1 | ||
| NM_001163436.2 | NP_001156908.1 | |||
| NM_001163437.2 | NP_001156909.1 | |||
| NM_001290768.1 | NP_001277697.1 | |||
| NM_033115.4 | NP_149106.2 | |||
| XM_011532417.2 | XP_011530719.1 | |||
| XM_017008846.1 | XP_016864335.1 | |||
| XM_017008847.1 | XP_016864336.1 | |||
| XM_017008848.1 | XP_016864337.1 | |||
| XM_017008849.1 | XP_016864338.1 | |||
| AB449876.1 | ||||
| AF161420.1 | AAF28980.1 | |||
| AK074305.1 | BAB85045.1 | |||
| AK098157.1 | BAC05244.1 | |||
| AK315571.1 | ||||
| BC009208.2 | AAH09208.2 | |||
| BC020853.2 | AAH20853.2 | |||
| BC068496.1 | AAH68496.1 | |||
| BX647851.1 | CAI46102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511247 | Chr.4:106101545 - 106144063 on Build GRCh38 | Gain |
|
| esv3601673 | Chr.4:106102808 - 106127774 on Build GRCh38 | Gain |
|
| nsv4450 | Chr.4:106116666 - 106161512 on Build GRCh38 | Deletion |
|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map