Assay Details
Target Gene Details
Entrez Gene ID: | 8671 |
Gene Name: | solute carrier family 4 member 4 |
Gene Aliases: |
HNBC1, KNBC, NBC1, NBC2, NBCe1-A, SLC4A5, hhNMC, kNBC1, pNBC |
Location: |
Chr.4:71187286-71572087 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC4A4 | NM_001098484.2 | NP_001091954.1 | ||
NM_001134742.1 | NP_001128214.1 | |||
AF011390.1 | AAC39840.1 | |||
AF157492.1 | AAF80343.1 | |||
AK307594.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
