Assay Details
Target Gene Details
Entrez Gene ID: | 4886 |
Gene Name: | neuropeptide Y receptor Y1 |
Gene Aliases: |
NPY1-R, NPYR |
Location: |
Chr.4:163323962-163344847 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY1R | XM_005263031.3 | XP_005263088.1 |
Target Gene Details
Entrez Gene ID: | 4889 |
Gene Name: | neuropeptide Y receptor Y5 |
Gene Aliases: |
NPY5-R, NPYR5, NPYY5-R |
Location: |
Chr.4:163336968-163352277 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY5R | XM_005263038.3 | 1 | 2872 | XP_005263095.1 |
| XM_011532015.2 | 1 | 2872 | XP_011530317.1 | |
| XM_011532017.2 | 1 | 2872 | XP_011530319.1 | |
| XM_017008256.1 | 1 | 2872 | XP_016863745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map