Assay Details
Target Gene Details
Entrez Gene ID: | 101927186 |
Gene Name: | adhesion G protein-coupled receptor L3 antisense RNA 1 |
Gene Aliases: |
LPHN3-AS1 |
Location: |
Chr.4:62071755-62161765 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL3-AS1 | NR_110595.1 | |||
| DB518756.1 |

Genomic Map