Assay Details
Target Gene Details
Entrez Gene ID: | 105378098 |
Gene Name: | uncharacterized LOC105378098 |
Gene Aliases: |
- |
Location: |
Chr.6:161470977-161484045 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105378098 | NR_134593.1 | |||
| NR_134594.1 | ||||
| NR_134595.1 | ||||
| DB041402.1 | ||||
| DB045952.1 | ||||
| DB083190.1 |
Target Gene Details
Entrez Gene ID: | 5071 |
Gene Name: | parkin RBR E3 ubiquitin protein ligase |
Gene Aliases: |
AR-JP, LPRS2, PDJ, PRKN |
Location: |
Chr.6:161347417-162727802 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARK2 | NM_004562.2 | NP_004553.2 | ||
| NM_013987.2 | NP_054642.2 | |||
| NM_013988.2 | NP_054643.2 | |||
| XM_011535863.1 | XP_011534165.1 | |||
| XM_017010908.1 | XP_016866397.1 | |||
| XM_017010909.1 | XP_016866398.1 | |||
| AB009973.1 | BAA25751.1 | |||
| AB245403.1 | ||||
| AF381282.1 | AAM21457.1 | |||
| AF381283.1 | AAM21458.1 | |||
| AF381284.1 | AAM21459.1 | |||
| AF381286.1 | AAM21461.1 | |||
| AK292590.1 | ||||
| AK294684.1 | ||||
| EF375726.1 | ||||
| GU345837.1 | ||||
| GU345838.1 | ||||
| GU345839.1 | ||||
| GU345840.1 | ||||
| GU357501.1 | ||||
| GU357502.1 | ||||
| GU361466.1 | ||||
| GU361467.1 | ||||
| GU361468.1 | ||||
| GU361469.1 | ||||
| GU361470.1 | ||||
| GU361471.1 | ||||
| KC357595.1 | ||||
| KC774171.1 | ||||
| KJ534911.1 | ||||
| KJ534912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830854 | Chr.6:161318894 - 161506483 on Build GRCh38 | Gain |
|
| nsv1031234 | Chr.6:161255736 - 161829694 on Build GRCh38 | Gain |
|
| esv3890978 | Chr.6:161452979 - 161535643 on Build GRCh38 | Loss |
|
| nsv1033603 | Chr.6:161423234 - 161580404 on Build GRCh38 | Loss |
|
| nsv1026543 | Chr.6:160306836 - 161585804 on Build GRCh38 | Gain |
|
| esv3890977 | Chr.6:161381113 - 161650603 on Build GRCh38 | Gain |
|
| nsv518931 | Chr.6:161372287 - 161985303 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76801163] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map