Assay Details
Target Gene Details
Entrez Gene ID: | 9844 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
CED-12, CED12, ELMO-1 |
Location: |
Chr.7:36852906-37449409 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1 | NM_001206480.2 | NP_001193409.1 | ||
| NM_001206482.1 | NP_001193411.1 | |||
| NM_014800.10 | NP_055615.8 | |||
| XM_005249919.2 | XP_005249976.1 | |||
| XM_006715805.1 | XP_006715868.1 | |||
| XM_011515654.1 | XP_011513956.1 | |||
| XM_017012838.1 | XP_016868327.1 | |||
| XM_017012839.1 | XP_016868328.1 | |||
| AF398885.1 | AAL14466.1 | |||
| AK126565.1 | BAC86597.1 | |||
| AK291784.1 | ||||
| AK296253.1 | ||||
| AK314349.1 | ||||
| AL136787.1 | CAB66721.1 | |||
| BC114341.2 | AAI14342.1 |
Target Gene Details
Entrez Gene ID: | 100861514 |
Gene Name: | ELMO1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:36997796-37013630 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1-AS1 | NR_104120.1 | |||
| DA058843.1 |
More Information
Set Membership: |
|

Genomic Map