Assay Details
Target Gene Details
Entrez Gene ID: | 51530 |
Gene Name: | zinc finger C3HC-type containing 1 |
Gene Aliases: |
HSPC216, NIPA |
Location: |
Chr.7:130018286-130051451 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3HC1 | NM_001282190.1 | NP_001269119.1 | ||
| NM_001282191.1 | NP_001269120.1 | |||
| NM_016478.4 | NP_057562.3 | |||
| XM_005250403.2 | XP_005250460.1 | |||
| XM_011516288.2 | XP_011514590.1 | |||
| XM_011516289.2 | XP_011514591.1 | |||
| XM_011516290.2 | XP_011514592.1 | |||
| XM_017012287.1 | XP_016867776.1 | |||
| XM_017012288.1 | XP_016867777.1 | |||
| AF151050.1 | AAF36136.1 | |||
| AJ537494.1 | CAD61161.1 | |||
| AK001317.1 | BAA91619.1 | |||
| AK022373.1 | BAB14024.1 | |||
| BC000190.1 | AAH00190.1 | |||
| BC011551.1 | AAH11551.1 | |||
| BC028917.1 | AAH28917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831129 | Chr.7:129885566 - 130062637 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map