Assay Details
Target Gene Details
Entrez Gene ID: | 1124 |
Gene Name: | chimerin 2 |
Gene Aliases: |
ARHGAP3, BCH, CHN2-3, RHOGAP3 |
Location: |
Chr.7:29146547-29514335 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHN2 | NM_001293069.1 | NP_001279998.1 | ||
| XM_011515105.2 | XP_011513407.1 | |||
| XM_011515106.2 | XP_011513408.1 | |||
| XM_017011721.1 | XP_016867210.1 | |||
| AK294271.1 | ||||
| DA190643.1 | ||||
| GQ924106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612685 | Chr.7:28997136 - 29209454 on Build GRCh38 | Loss |
|
| nsv1019174 | Chr.7:29163105 - 29499710 on Build GRCh38 | Gain |
|
| esv2761314 | Chr.7:29151010 - 29498723 on Build GRCh38 | Gain |
|
| nsv5680 | Chr.7:29161353 - 29194769 on Build GRCh38 | Insertion |
|
| dgv11248n54 | Chr.7:29169736 - 29497091 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map