Assay Details
Target Gene Details
Entrez Gene ID: | 3082 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
DFNB39, F-TCF, HGFB, HPTA, SF |
Location: |
Chr.7:81699006-81770438 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HGF | NM_000601.5 | 18 | 4152 | NP_000592.3 |
| NM_001010932.2 | 18 | 4137 | NP_001010932.1 | |
| XM_006715956.2 | 19 | 4115 | XP_006716019.1 | |
| XM_011516115.2 | 19 | 4100 | XP_011514417.1 | |
| X16323.1 | 18 | 4059 | CAA34387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576398 | Chr.7:81685626 - 81732874 on Build GRCh38 | Gain |
|
| nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map