Assay Details
Target Gene Details
Entrez Gene ID: | 5444 |
Gene Name: | paraoxonase 1 |
Gene Aliases: |
ESA, MVCD5, PON |
Location: |
Chr.7:95298357-95324572 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PON1 | NM_000446.5 | NP_000437.3 | ||
| AK314027.1 | ||||
| BC074719.2 | AAH74719.1 | |||
| BJ993711.1 | ||||
| CB122285.1 | ||||
| D84371.1 | BAA12327.1 | |||
| M63012.1 | AAB59538.1 | |||
| M63013.1 | AAA60142.1 | |||
| M63014.1 | AAA60143.1 | |||
| S64615.1 | AAB27714.2 | |||
| U53784.1 | AAA97957.1 | |||
| Z70723.1 | CAA94728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015973 | Chr.7:95312103 - 95339029 on Build GRCh38 | Loss |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
| nsv528555 | Chr.7:95311275 - 95329316 on Build GRCh38 | Loss |
|
| nsv5851 | Chr.7:95308446 - 95339854 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map