Assay Details
Target Gene Details
Entrez Gene ID: | 253012 |
Gene Name: | HEPACAM family member 2 |
Gene Aliases: |
MIKI |
Location: |
Chr.7:93188586-93232297 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEPACAM2 | NM_001039372.2 | NP_001034461.1 | ||
| NM_001288804.1 | NP_001275733.1 | |||
| NM_001288810.1 | NP_001275739.1 | |||
| NM_198151.2 | NP_937794.1 | |||
| XM_011516000.2 | XP_011514302.1 | |||
| XM_011516001.1 | XP_011514303.1 | |||
| AK096002.1 | ||||
| AK298361.1 | ||||
| AY358345.1 | AAQ88711.1 | |||
| BC136300.1 | ||||
| BC136301.1 | ||||
| BC139906.1 | ||||
| DA426282.1 |
More Information
Set Membership: |
|

Genomic Map