Assay Details
Target Gene Details
| Entrez Gene ID: | 100505854 | 
| Gene Name: | Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding) | 
| Gene Aliases: | RSBN1L-AS1 | 
| Location: | Chr.7:77657660-77697345 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| APTR | NR_134251.1 | |||
| NR_134253.1 | ||||
| NR_134254.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3613804 | Chr.7:77676857 - 77687048 on Build GRCh38 | Loss |  APTR | 
| dgv6510n100 | Chr.7:77643042 - 77681947 on Build GRCh38 | Loss |  APTR | 
| esv2661869 | Chr.7:77676828 - 77687192 on Build GRCh38 | Deletion |  APTR | 
| nsv831039 | Chr.7:77564611 - 77722887 on Build GRCh38 | Gain |  APTR  PTPN12  RSBN1L | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic