Assay Details
Target Gene Details
Entrez Gene ID: | 5445 |
Gene Name: | paraoxonase 2 |
Gene Aliases: |
- |
Location: |
Chr.7:95404862-95435072 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PON2 | NM_000305.2 | NP_000296.2 | ||
| NM_001018161.1 | NP_001018171.1 | |||
| XM_005250453.1 | XP_005250510.1 | |||
| XM_017012357.1 | XP_016867846.1 | |||
| XM_017012358.1 | XP_016867847.1 | |||
| AB102891.1 | BAD89420.1 | |||
| AF001601.1 | AAC27944.1 | |||
| AF001602.1 | AAC27945.1 | |||
| AK291103.1 | ||||
| AK296029.1 | ||||
| AK296636.1 | ||||
| AK301313.1 | ||||
| AK308065.1 | ||||
| AK315209.1 | ||||
| BC040010.1 | AAH40010.1 | |||
| BC046160.1 | AAH46160.1 | |||
| L48513.1 | AAC41995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map