Assay Details
Target Gene Details
Entrez Gene ID: | 27102 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 1 |
Gene Aliases: |
HCR, HRI |
Location: |
Chr.7:6022247-6059229 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK1 | NM_001134335.1 | NP_001127807.1 | ||
| NM_014413.3 | NP_055228.2 | |||
| AB037790.1 | ||||
| AF085994.1 | ||||
| AF100784.1 | AAP97223.1 | |||
| AF147094.1 | AAF66736.1 | |||
| AF183414.1 | AAG09683.1 | |||
| AF255050.1 | AAF70289.1 | |||
| AK075192.1 | BAC11461.1 | |||
| AK290327.1 | ||||
| AK295712.1 | ||||
| AL136563.1 | CAB66498.1 | |||
| AL834494.1 | CAD39152.1 | |||
| BC006524.2 | AAH06524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034916 | Chr.7:5895149 - 6119339 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv605998 | Chr.7:5995434 - 6062403 on Build GRCh38 | Loss |
|
| nsv1018388 | Chr.7:5999082 - 6091538 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map