Assay Details
Target Gene Details
Entrez Gene ID: | 8662 |
Gene Name: | eukaryotic translation initiation factor 3 subunit B |
Gene Aliases: |
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1 |
Location: |
Chr.7:2354540-2380745 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3B | NM_001037283.1 | NP_001032360.1 | ||
| NM_003751.3 | NP_003742.2 | |||
| XM_011515599.1 | XP_011513901.1 | |||
| XM_011515600.1 | XP_011513902.1 | |||
| XM_011515601.1 | XP_011513903.1 | |||
| XM_017012752.1 | XP_016868241.1 | |||
| XM_017012753.1 | XP_016868242.1 | |||
| AK300967.1 | ||||
| AK302058.1 | ||||
| BC001173.1 | AAH01173.1 | |||
| BC110865.1 | AAI10866.1 | |||
| BG325094.1 | ||||
| U62583.1 | AAB42010.1 | |||
| U78525.1 | AAC99479.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73043300] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map