Assay Details
Target Gene Details
Entrez Gene ID: | 100505881 |
Gene Name: | MAGI2 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.7:79452957-79471208 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGI2-AS3 | NR_038343.2 | 4 | 5755 | |
| NR_038344.1 | ||||
| NR_038345.1 | ||||
| NR_038346.1 | ||||
| AB371456.1 | ||||
| BI521274.1 | ||||
| BI868593.1 | ||||
| CN311982.1 | ||||
| CR749529.1 | 3 | 5726 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607659 | Chr.7:78863326 - 79854164 on Build GRCh38 | Gain |
|
| esv2764028 | Chr.7:79245424 - 79483314 on Build GRCh38 | Gain+Loss |
|

Genomic Map