Assay Details
Target Gene Details
Entrez Gene ID: | 57180 |
Gene Name: | ARP3 actin related protein 3 homolog B |
Gene Aliases: |
ARP11, ARP3BETA |
Location: |
Chr.7:152759749-153070979 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR3B | XM_006716065.2 | XP_006716128.1 | ||
| XM_017012460.1 | XP_016867949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034937 | Chr.7:152869077 - 153157810 on Build GRCh38 | Gain |
|
| nsv6008 | Chr.7:152866185 - 152900172 on Build GRCh38 | Deletion |
|
| esv3615531 | Chr.7:152820301 - 152885417 on Build GRCh38 | Gain |
|
| nsv508491 | Chr.7:152861530 - 152891147 on Build GRCh38 | Deletion |
|
| esv3615532 | Chr.7:152837608 - 152876169 on Build GRCh38 | Gain |
|
| nsv831193 | Chr.7:152656459 - 152895593 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76330036] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map