Assay Details
Target Gene Details
Entrez Gene ID: | 1644 |
Gene Name: | dopa decarboxylase |
Gene Aliases: |
AADC |
Location: |
Chr.7:50458436-50565460 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDC | NM_000790.3 | NP_000781.1 | ||
| NM_001082971.1 | NP_001076440.1 | |||
| NM_001242886.1 | NP_001229815.1 | |||
| NM_001242887.1 | NP_001229816.1 | |||
| NM_001242888.1 | NP_001229817.1 | |||
| NM_001242889.1 | NP_001229818.1 | |||
| XM_005271745.4 | XP_005271802.1 | |||
| XM_011515161.2 | XP_011513463.2 | |||
| AK298321.1 | ||||
| AK298392.1 | ||||
| BC000485.1 | AAH00485.1 | |||
| BC008366.1 | AAH08366.1 | |||
| BT007009.1 | AAP35655.1 | |||
| CD013934.1 | ||||
| CD013935.1 | ||||
| CD013936.1 | ||||
| CD013937.1 | ||||
| CD013938.1 | ||||
| CR456724.1 | CAG33005.1 | |||
| KU178021.1 | ||||
| KU178022.1 | ||||
| M76180.1 | AAA58437.1 | |||
| M88700.1 | AAA20894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016665 | Chr.7:50384854 - 51336827 on Build GRCh38 | Gain |
|
| nsv1026081 | Chr.7:50458202 - 50470486 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73695682] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map