Assay Details
Target Gene Details
Entrez Gene ID: | 375611 |
Gene Name: | solute carrier family 26 member 5 |
Gene Aliases: |
DFNB61, PRES |
Location: |
Chr.7:103352598-103446177 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A5 | NM_001321787.1 | NP_001308716.1 | ||
| NM_206883.2 | NP_996766.1 | |||
| NM_206884.2 | NP_996767.1 | |||
| NM_206885.2 | NP_996768.1 | |||
| NR_135802.1 | ||||
| XM_011516170.2 | 20 | 7094 | XP_011514472.1 | |
| AY256823.1 | AAP31532.1 | |||
| AY256824.1 | AAP31533.1 | |||
| AY256825.1 | AAP31534.1 |
More Information
Set Membership: |
|

Genomic Map