Assay Details
Target Gene Details
Entrez Gene ID: | 231 |
Gene Name: | aldo-keto reductase family 1 member B |
Gene Aliases: |
ADR, ALDR1, ALR2, AR |
Location: |
Chr.7:134442352-134459136 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1B1 | NM_001628.2 | NP_001619.1 | ||
| XM_005250234.3 | XP_005250291.1 | |||
| AB209796.1 | BAD93033.1 | |||
| AF328729.1 | AAN09721.1 | |||
| AK313439.1 | ||||
| AL833410.1 | 1 | 1593 | ||
| BC000260.1 | AAH00260.1 | |||
| BC005387.1 | AAH05387.1 | |||
| BC010391.1 | AAH10391.1 | |||
| BT019859.1 | AAV38662.1 | |||
| CR450351.1 | CAG29347.1 | |||
| CR542203.1 | CAG47000.1 | |||
| J04795.1 | AAA51713.1 | |||
| J05017.1 | AAA51714.1 | |||
| J05474.1 | AAA51715.1 | |||
| M34720.1 | AAA35560.1 | |||
| X15414.1 | CAA33460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615093 | Chr.7:134265041 - 134524737 on Build GRCh38 | Gain |
|
| esv3615084 | Chr.7:133940538 - 134524796 on Build GRCh38 | Gain |
|
| nsv1020759 | Chr.7:133928362 - 134443062 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map