Assay Details
Target Gene Details
Entrez Gene ID: | 6677 |
Gene Name: | sperm adhesion molecule 1 |
Gene Aliases: |
HEL-S-96n, HYA1, HYAL1, HYAL3, HYAL5, PH-20, PH20, SPAG15 |
Location: |
Chr.7:123925232-123971410 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAM1 | NM_001174044.1 | NP_001167515.1 | ||
| NM_001174045.1 | NP_001167516.1 | |||
| NM_001174046.1 | NP_001167517.1 | |||
| NM_003117.4 | NP_003108.2 | |||
| NM_153189.2 | NP_694859.1 | |||
| AK292229.1 | ||||
| AY920278.1 | AAX13985.1 | |||
| AY920279.1 | AAX13986.1 | |||
| AY920280.1 | AAX13987.1 | |||
| AY920281.1 | AAX13988.1 | |||
| BC026163.1 | AAH26163.1 | |||
| EU794686.1 | ||||
| L13781.1 | ||||
| S67798.1 | AAC60607.2 | |||
| X84347.1 | CAA59086.1 |
More Information
Set Membership: |
|

Genomic Map