Assay Details
Target Gene Details
Entrez Gene ID: | 93986 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
CAGH44, SPCH1, TNRC10 |
Location: |
Chr.7:114086310-114693772 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP2 | NR_033766.1 | |||
XM_017012801.1 | XP_016868290.1 | |||
AF454830.1 | AAM60761.1 | |||
AK131266.1 | BAD18444.1 | |||
CR749236.1 | CAH18092.1 | |||
DA400243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv608231 | Chr.7:114067732 - 114419101 on Build GRCh38 | Loss |
![]() |
nsv608230 | Chr.7:113941914 - 114186383 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
