Assay Details
Target Gene Details
Entrez Gene ID: | 100996928 |
Gene Name: | C7orf55-LUC7L2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:139341132-139423457 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf55-LUC7L2 | NM_001244584.2 | NP_001231513.1 | ||
| AK297642.1 |
Target Gene Details
Entrez Gene ID: | 51631 |
Gene Name: | LUC7 like 2, pre-mRNA splicing factor |
Gene Aliases: |
CGI-59, CGI-74, LUC7B2 |
Location: |
Chr.7:139340359-139423457 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUC7L2 | NM_001244585.1 | NP_001231514.1 | ||
| NM_001270643.1 | NP_001257572.1 | |||
| NM_016019.4 | NP_057103.2 | |||
| AF151817.1 | AAD34054.1 | |||
| AF151832.1 | AAD34069.1 | |||
| AK001476.1 | BAA91713.1 | |||
| AK001519.1 | BAA91737.1 | |||
| AK022895.1 | BAB14297.1 | |||
| AK093864.1 | ||||
| AK298166.1 | ||||
| BC017163.2 | AAH17163.1 | |||
| BC042625.1 | AAH42625.1 | |||
| BC050708.2 | AAH50708.1 | |||
| BC056886.1 | AAH56886.1 | |||
| DA342563.1 | ||||
| DB086483.1 | ||||
| HY018775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map