Assay Details
Target Gene Details
Entrez Gene ID: | 27102 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 1 |
Gene Aliases: |
HCR, HRI |
Location: |
Chr.7:6022247-6059229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2AK1 | NM_001134335.1 | NP_001127807.1 | ||
NM_014413.3 | NP_055228.2 | |||
AB037790.1 | ||||
AF100784.1 | AAP97223.1 | |||
AF147094.1 | AAF66736.1 | |||
AF183414.1 | AAG09683.1 | |||
AF255050.1 | AAF70289.1 | |||
AK075192.1 | BAC11461.1 | |||
AK290327.1 | ||||
AK295712.1 | ||||
AL136563.1 | CAB66498.1 | |||
AL834494.1 | CAD39152.1 | |||
BC006524.2 | AAH06524.1 | |||
DC341983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1034916 | Chr.7:5895149 - 6119339 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv605998 | Chr.7:5995434 - 6062403 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv1018388 | Chr.7:5999082 - 6091538 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
