Assay Details
Target Gene Details
Entrez Gene ID: | 56302 |
Gene Name: | transient receptor potential cation channel subfamily V member 5 |
Gene Aliases: |
CAT2, ECAC1, OTRPC3 |
Location: |
Chr.7:142908101-142933723 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV5 | NM_019841.6 | NP_062815.3 | ||
| AF304464.1 | AAL04015.1 | |||
| AJ271207.2 | CAB96365.2 | |||
| AJ487965.2 | CAD32312.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831168 | Chr.7:142830591 - 143025918 on Build GRCh38 | Gain |
|
| esv2759569 | Chr.7:142434958 - 143027060 on Build GRCh38 | Loss |
|
| nsv526015 | Chr.7:142864981 - 142944120 on Build GRCh38 | Gain |
|
| nsv520735 | Chr.7:142865498 - 142944120 on Build GRCh38 | Loss |
|
| nsv1030700 | Chr.7:142867398 - 143010429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map