Assay Details
Target Gene Details
Entrez Gene ID: | 800 |
Gene Name: | caldesmon 1 |
Gene Aliases: |
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22 |
Location: |
Chr.7:134779413-134970729 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALD1 | NM_004342.6 | NP_004333.1 | ||
| NM_033138.3 | NP_149129.2 | |||
| NM_033157.3 | NP_149347.2 | |||
| AF247820.3 | AAF69498.3 | |||
| AK289686.1 | ||||
| AK298524.1 | ||||
| AK304729.1 | ||||
| BC040354.1 | AAH40354.1 | |||
| CD359921.1 | ||||
| DB090561.1 | ||||
| M83216.1 | AAA58420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764025 | Chr.7:134799276 - 134975815 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map