Assay Details
Target Gene Details
Entrez Gene ID: | 2146 |
Gene Name: | enhancer of zeste 2 polycomb repressive complex 2 subunit |
Gene Aliases: |
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2 |
Location: |
Chr.7:148807372-148884662 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522305 | Chr.7:148771525 - 148811105 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map