Assay Details
Target Gene Details
Entrez Gene ID: | 23072 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
NEDL1 |
Location: |
Chr.7:43112580-43566001 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Target Gene Details
Entrez Gene ID: | 100506895 |
Gene Name: | uncharacterized LOC100506895 |
Gene Aliases: |
- |
Location: |
Chr.7:43508728-43522542 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506895 | NR_038276.1 | 3 | 2344 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612994 | Chr.7:43438723 - 43515026 on Build GRCh38 | Gain |
|
| esv3612993 | Chr.7:43438543 - 43609526 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74368380] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map