Assay Details
Target Gene Details
Entrez Gene ID: | 83988 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
- |
Location: |
Chr.8:101686542-102124907 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCALD | NM_001040624.1 | NP_001035714.1 | ||
| NM_001040625.1 | NP_001035715.1 | |||
| NM_001040626.1 | NP_001035716.1 | |||
| NM_001040627.1 | NP_001035717.1 | |||
| NM_001040628.1 | NP_001035718.1 | |||
| NM_001040629.1 | NP_001035719.1 | |||
| NM_001040630.1 | NP_001035720.1 | |||
| NM_032041.2 | NP_114430.2 | |||
| XM_011517332.2 | XP_011515634.1 | |||
| XM_011517333.2 | XP_011515635.1 | |||
| XM_011517334.2 | XP_011515636.1 | |||
| XM_011517335.2 | XP_011515637.1 | |||
| XM_017013901.1 | XP_016869390.1 | |||
| XM_017013902.1 | XP_016869391.1 | |||
| AB209015.1 | BAD92252.1 | |||
| AF251061.1 | AAK34951.1 | |||
| AK055073.1 | ||||
| AK055097.1 | 1 | 302 | ||
| AK096515.1 | ||||
| AK314523.1 | ||||
| AL136612.1 | CAB66547.1 | |||
| BC036098.1 | AAH36098.1 | |||
| BC063428.1 | AAH63428.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115241089] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map