Assay Details
Target Gene Details
Entrez Gene ID: | 1978 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 1 |
Gene Aliases: |
4E-BP1, 4EBP1, BP-1, PHAS-I |
Location: |
Chr.8:38030502-38060365 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4EBP1 | NM_004095.3 | NP_004086.1 | ||
| AB044548.1 | BAB18650.1 | |||
| AK312011.1 | ||||
| BC004459.1 | AAH04459.1 | |||
| BC058073.1 | AAH58073.1 | |||
| BF964628.1 | ||||
| BT007162.1 | AAP35826.1 | |||
| CR456769.1 | CAG33050.1 | |||
| L36055.1 | AAA62269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616914 | Chr.8:38051747 - 38056815 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map