Assay Details
Target Gene Details
Entrez Gene ID: | 286097 |
Gene Name: | mitochondrial calcium uptake family member 3 |
Gene Aliases: |
EFHA2 |
Location: |
Chr.8:17027238-17133069 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICU3 | NM_181723.2 | NP_859074.1 | ||
| XM_005273477.2 | XP_005273534.1 | |||
| XM_006716328.2 | XP_006716391.1 | |||
| XM_006716329.2 | XP_006716392.1 | |||
| XM_006716330.2 | XP_006716393.1 | |||
| XM_006716333.3 | XP_006716396.1 | |||
| XM_011544507.2 | XP_011542809.1 | |||
| XM_011544508.2 | XP_011542810.1 | |||
| XM_017013336.1 | XP_016868825.1 | |||
| XM_017013337.1 | XP_016868826.1 | |||
| XM_017013338.1 | XP_016868827.1 | |||
| XM_017013339.1 | XP_016868828.1 | |||
| XM_017013340.1 | XP_016868829.1 | |||
| XM_017013341.1 | XP_016868830.1 | |||
| XM_017013342.1 | XP_016868831.1 | |||
| XM_017013343.1 | XP_016868832.1 | |||
| BC032868.1 | AAH32868.1 | |||
| BC045553.1 | AAH45553.1 | |||
| BI546243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831243 | Chr.8:16899975 - 17111869 on Build GRCh38 | Gain |
|
| esv2759599 | Chr.8:16330049 - 17108753 on Build GRCh38 | Gain |
|
| nsv1018645 | Chr.8:16697556 - 17111822 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79898401] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map