Assay Details
Target Gene Details
Entrez Gene ID: | 210 |
Gene Name: | aminolevulinate dehydratase |
Gene Aliases: |
ALADH, PBGS |
Location: |
Chr.9:113386312-113401338 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALAD | NM_000031.5 | NP_000022.3 | ||
| NM_001003945.2 | NP_001003945.1 | |||
| NM_001317745.1 | NP_001304674.1 | |||
| XM_011518364.2 | XP_011516666.1 | |||
| AK131490.1 | BAD18635.1 | |||
| AK290490.1 | ||||
| AK295945.1 | ||||
| AK312552.1 | ||||
| BC000977.3 | AAH00977.3 | |||
| BX494400.1 | ||||
| M13928.1 | AAA51687.1 | |||
| S99468.1 | AAC60581.1 | |||
| S99471.1 | AAC60582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621540 | Chr.9:113279513 - 113421563 on Build GRCh38 | Gain |
|
| nsv825053 | Chr.9:113388707 - 113396712 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map