Assay Details
Target Gene Details
Entrez Gene ID: | 2776 |
Gene Name: | G protein subunit alpha q |
Gene Aliases: |
CMC1, G-ALPHA-q, GAQ, SWS |
Location: |
Chr.9:77716274-78031449 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAQ | NM_002072.4 | NP_002063.2 | ||
| XM_017014628.1 | XP_016870117.1 | |||
| AF011496.1 | AAB64301.1 | |||
| AF329284.1 | AAG61117.1 | |||
| AF493896.1 | AAM12610.1 | |||
| AK298579.1 | ||||
| BC057777.1 | AAH57777.1 | |||
| BC067850.1 | ||||
| BC069520.1 | AAH69520.1 | |||
| BC075096.2 | AAH75096.1 | |||
| BC075097.2 | AAH75097.1 | |||
| JN184778.1 | ||||
| L40629.1 | AAA99950.1 | |||
| L76256.1 | AAB39498.1 | |||
| U40038.1 | AAC50363.1 | |||
| U43083.1 | AAB06875.1 |
More Information
Set Membership: |
|

Genomic Map