Assay Details
Target Gene Details
Entrez Gene ID: | 221035 |
Gene Name: | receptor accessory protein 3 |
Gene Aliases: |
C10orf74, Yip2b |
Location: |
Chr.10:63521353-63625123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
REEP3 | NM_001001330.2 | NP_001001330.1 | ||
XM_011539501.2 | XP_011537803.1 | |||
XM_017015896.1 | XP_016871385.1 | |||
AK289976.1 | ||||
AY562241.1 | AAT70686.1 | |||
BC010040.2 | AAH10040.1 | |||
BC057832.1 | AAH57832.1 | |||
BC068557.1 | AAH68557.1 | |||
KJ534927.1 | ||||
KJ535017.1 | ||||
KJ535031.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551184 | Chr.10:63501424 - 63719655 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
