Assay Details
Target Gene Details
Entrez Gene ID: | 1791 |
Gene Name: | DNA nucleotidylexotransferase |
Gene Aliases: |
TDT |
Location: |
Chr.10:96304328-96338564 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNTT | NM_001017520.1 | NP_001017520.1 | ||
NM_004088.3 | NP_004079.3 | |||
AB046378.1 | BAB72001.1 | |||
AK130771.1 | ||||
AK223317.1 | BAD97037.1 | |||
AK313615.1 | ||||
BC012920.1 | AAH12920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831950 | Chr.10:96321215 - 96485677 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2761620 | Chr.10:96287288 - 96329992 on Build GRCh38 | Gain |
![]() |
nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
